Human Recombinant sIL-6Ra (from CHO Cells)
IL-6 mediates its biological effects through the type I IL-6 receptor system that consists of two chains, IL-6Rα and gp130. While the IL-6Rα chain is the binding component specific to IL-6, the gp130 chain only transmits signals of IL-6 when bound to IL-6Rα. The gp130 can also transmit signals from LIF, OSM, CNTF, IL-11 and CT-1 in conjunction with other receptor subunits. The low-affinity binding site for IL-6 is composed of IL-6Rα alone. IL-6Rα is expressed in a wide range of cells, including T cells, fibroblasts and macrophages. Soluble IL-6Rα, which consists of only the extracellular domain of the IL-6Rα chain, acts as an agonist of IL-6 activity at low concentrations. PeproTech's
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:CHO cells
- Species:Human
- Storage Conditions:Store at –20 °C prior to reconstitution.
- Endotoxin Content:>0.1 ng/μg
- Biological Activity:Biological Activity tests can vary. Please refer to the Product CoA for information.
- Form:Lyophilized
- Gene ID:3570 P08887
- Reconstitution Instructions:Reconstitution instructions can vary. Please refer to the product CoA for info.
- Protease-free:Y
- Protein Synonyms:soluble Interleukin-6Rα|soluble IL-6 Receptor alpha|CD126
- Protein/Peptide Name:sIL-6Ra
- Purity:0.95
- Molecular Weight:37.9 kDa
- Sequence:LAPRRCPAQE VARGVLTSLP GDSVTLTCPG VEPEDNATVH WVLRKPAAGS HPSRWAGMGR RLLLRSVQLH DSGNYSCYRA GRPAGTVHLL VDVPPEEPQL SCFRKSPLSN VVCEWGPRST PSLTTKAVLL VRKFQNSPAE DFQEPCQYSQ ESQKFSCQLA VPEGDSSFYI VSMCVASSVG SKFSKTQTFQ GCGILQPDPP ANITVTAVAR NPRWLSVTWQ DPHSWNSSFY RLRFELRYRA ERSKTFTTWM VKDLQHHCVI HDAWSGLRHV VQLRAQEEFG QGEWSEWSPE AMGTPWTESR SPPAENEVST PMQALTTNKD DDNILFRDSA NATSLPVQD
- Endotoxin Level:Low
- Formulation:Formulation can vary. Please refer to product CoA for information.
- Shipping Temperature:Ambient
Frequently Bought Together