About this item
C1 inhibitor is a member of the serpin family of structurally related proteins, and is the primary regulator of the immune complement system. C1 inhibitor is a protease inhibitor that functions to inhibit the complement system in order to prevent over-activation or spontaneous activation. Inhibition is achieved by binding to and irreversibly inhibiting the C1r and C1s proteases of the C1 complex, which has the effect of shutting down all subsequent downstream events in the complement activation cascade. C1 inhibitor can also inhibit various other proteases, including Kallikrein, Factor XIa, and Factor XIIa. Deficiencies in C1 inhibitor are the primary cause of hereditary angioedema (HAE, hereditary angioneurotic edema), a disease characterized by edema in the respiratory and gastrointestinal tracts. In certain clinical situations, the direct administration of C1 inhibitor can be used to treat HAE and certain other conditions. Recombinant Human C1 Inhibitor is a highly glycosylated glycoprotein containing 478 amino acid residues (52.8kDa), corresponding to amino acids 23 – 500 of the C1 inhibitor precursor, and is fully functional in its ability to inhibit the C1 complex. Glycosylated C1 Inhibitor migrates at an apparent molecular weight of approximately 80-90 kDa by SDS PAGE analysis under reducing conditions.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:CHO cells
- Species:Human
- Storage Conditions:Store at –20 °C prior to reconstitution.
- Endotoxin Content:>0.1 ng/μg
- Biological Activity:Biological Activity tests can vary. Please refer to the Product CoA for information.
- Form:Lyophilized
- Gene ID:1493 P16410
- Reconstitution Instructions:Reconstitution instructions can vary. Please refer to the product CoA for info.
- Protease-free:Y
- Protein Synonyms:Cytotoxic T-lymphocyte-associated antigen 4|CD152
- Protein/Peptide Name:CTLA-4
- Purity:0.98
- Molecular Weight:78.7 kDa
- Sequence:MHVAQPAVVL ASSRGIASFV CEYASPGKAT EVRVTVLRQA DSQVTEVCAA TYMMGNELTF LDDSICTGTS SGNQVNLTIQ GLRAMDTGLY ICKVELMYPP PYYLGIGNGT QIYVIDPEPC PDSDGGPKSC DKTHTCPPCP APELLGGPSV FLFPPKPKDT LMISRTPEVT CVVVDVSHED PEVKFNWYVD GVEVHNAKTK PREEQYNSTY RVVSVLTVLH QDWLNGKEYK CKVSNKALPA PIEKTISKAK GQPREPQVYT LPPSRDELTK NQVSLTCLVK GFYPSDIAVE WESNGQPENN YKTTPPVLDS DGSFFLYSKL TVDKSRWQQG NVFSCSVMHE ALHNHYTQKS LSLSPGK
- Endotoxin Level:Low
- Formulation:Formulation can vary. Please refer to product CoA for information.
- Shipping Temperature:Ambient