Human IL-1α Standard TMB ELISA Development Kit, PeproTech, Inc.
PeproTech's Human IL-1α ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant human IL-1α in a sandwich ELISA format.
Human IL-1α ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant human IL-1α in a sandwich ELISA format. Using the ELISA protocol, the recommended microplates, reagents and solutions, the components supplied in this kit are sufficient to assay human IL-1α in approximately 1000 ELISA plate wells.
- Assay Type (ELISA with LOV):Sandwich
- Primary antibody reactivity:Human
- Target protein:IL-1α
Frequently Bought Together