Rat TNF-α Standard ABTS ELISA Development Kit, PeproTech
Supplier: Peprotech
Certificates
About this item
PeproTech's Rat TNF-α ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant rat TNF-α in a sandwich ELISA format.
- Easy-to-use
- Optimized reagents for singleplex protein detection by solid phase sandwich enzyme-linked immunosorbent assay (ELISA)
Rat TNF-α ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant rat TNF-α in a sandwich ELISA format. Using the ELISA protocol, the recommended microplates, reagents and solutions, the components supplied in this kit are sufficient to assay rat TNF-α in approximately 1000 ELISA plate wells.
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Target protein:TNF-α
Specifications
Frequently Bought Together