Rat EGF Standard ABTS ELISA Development Kit, PeproTech
Supplier: Peprotech
Certificates
About this item
PeproTech's Rat EGF ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant rat EGF in a sandwich ELISA format.
- Easy-to-use
- Optimized reagents for singleplex protein detection by solid phase sandwich enzyme-linked immunosorbent assay (ELISA)
Rat EGF ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant rat EGF in a sandwich ELISA format. Using the ELISA protocol, the recommended microplates, reagents and solutions, the components supplied in this kit are sufficient to assay rat EGF in approximately 1000 ELISA plate wells.
Frequently Bought Together