Human TL-1A Standard ABTS ELISA Development Kit, PeproTech, Inc.
Supplier: Peprotech
Certificates
About this item
PeproTech's Human TL-1A ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant human TL-1A in a sandwich ELISA format.
Human TL-1A ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant human TL-1A in a sandwich ELISA format. Using the ELISA protocol, the recommended microplates, reagents and solutions, the components supplied in this kit are sufficient to assay human TL-1A in approximately 1000 ELISA plate wells.
Specifications
- Primary antibody reactivity:Human