Human Resistin Standard ABTS ELISA Development Kit, PeproTech
Supplier: Peprotech
Certificates
About this item
PeproTech's Human Resistin ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant human Resistin in a sandwich ELISA format.
- Easy-to-use
- Optimized reagents for singleplex protein detection by solid phase sandwich enzyme-linked immunosorbent assay (ELISA)
Human Resistin ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant human Resistin in a sandwich ELISA format. Using the ELISA protocol, the recommended microplates, reagents and solutions, the components supplied in this kit are sufficient to assay human Resistin in approximately 1000 ELISA plate wells.
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Primary antibody reactivity:Human
- Target protein:Resistin
Specifications
Frequently Bought Together