Human BD-3 Standard ABTS ELISA Development Kit, PeproTech
PeproTech's Human BD-3 ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant human BD-3 in a sandwich ELISA format.
- Easy-to-use
- Optimized reagents for singleplex protein detection by solid phase sandwich enzyme-linked immunosorbent assay (ELISA)
Human BD-3 ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant human BD-3 in a sandwich ELISA format. Using the ELISA protocol, the recommended microplates, reagents and solutions, the components supplied in this kit are sufficient to assay human BD-3 in approximately 1000 ELISA plate wells.
- Primary antibody reactivity:Human
Frequently Bought Together