Specifications
- Host:
- Primary antibody reactivity:Human
- Description:Human IL-7 ELISA Kit
- Size:Kit components sufficient to assay 10 plates
- Environmentally Preferable:
- Cat. No.:10777-650
- Supplier no.:900-K17
Specifications
About this item
PeproTech's Human IL-7 ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant human IL-7 in a sandwich ELISA format.
- Easy-to-use
- Optimized reagents for singleplex protein detection by solid phase sandwich enzyme-linked immunosorbent assay (ELISA)
Human IL-7 ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant human IL-7 in a sandwich ELISA format. Using the ELISA protocol, the recommended microplates, reagents and solutions, the components supplied in this kit are sufficient to assay human IL-7 in approximately 1000 ELISA plate wells.