Murine JE/MCP-1 (CCL2) Standard ABTS ELISA Development Kit, PeproTech
Supplier: Peprotech
Certificates
About this item
PeproTech's Murine JE/MCP-1 (CCL2) ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant murine JE/MCP-1 (CCL2) in a sandwich ELISA format.
- Easy-to-use
- Optimized reagents for singleplex protein detection by solid phase sandwich enzyme-linked immunosorbent assay (ELISA)
Murine JE/MCP-1 (CCL2) ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant murine JE/MCP-1 (CCL2) in a sandwich ELISA format. Using the ELISA protocol, the recommended microplates, reagents and solutions, the components supplied in this kit are sufficient to assay murine JE/MCP-1 (CCL2) approximately 1000 ELISA plate wells.
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Primary antibody reactivity:RatMouse
- Target protein:MCP-1 (CCL2)
Specifications
Frequently Bought Together