Orders LinkContactUsLinkComponent
UV Flashlight
UV Flashlight
UV flashlight, 365 nm, 3¼", ¾"
Catalog # 470304-018
Supplier #:  AISUVLT1
UV Flashlight
Catalog # 470304-018
UV Flashlight
UV Flashlight
UV flashlight, 365 nm, 3¼", ¾"
Price
$15.40
Log in for availability
Each
Product Details & Documents
Powerful UV light for mineral fluorescence.
  • Bright 365 nm single LED bulb
  • Rechargeable via a USB cord (included)
  • Illuminates a large area
This compact UV flashlight packs a lot of power in a small package. The UV flashlight is ideal for forensics or illuminating fluorescent and blacklight reactive substances. The long-life UV LED has a wavelength of 365 nm (UV-A). An attached lanyard allows secure handling throughout use, so it never falls to the ground. The rechargeable battery connects to any USB port for charging on the go. Charging cord is included, USB 2.0 Micro-B.
Caution: Do not run the flashlight for more than 8 continuous minutes.
Specifications
  • Ø:
    ¾"
  • Description:
    UV flashlight
  • Length:
    3¼"
  • Wavelength:
    365 nm