ContactUsLinkComponent
Hi-Level Bandage Scissors, OR-Grade, Sklar®
Hi-Level Bandage Scissors, OR-Grade, Sklar®
Catalog # 76191-762
Supplier:  Sklar
undefined
Hi-Level Bandage Scissors, OR-Grade, Sklar®
Catalog # 76191-762
Supplier:  Sklar
Supplier Number:  15-3354
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Length:
    5.5"
  • Material:
    Stainless steel
  • Curvature:
    Straight
  • Cat. No.:
    76191-762
  • Blade type:
    Sharp/Blunt
Specifications
About this item
All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
Supercut Sklarhone™ Hi-Level Bandage Scissors are non-ratcheted, finger ring scissors primarily used for removing tight bandages. The tip of the lower blade features a flattened blunt nodule that is intended to slide between bandages and skin without harming the skin. Supercut Sklarhone™ scissors combine one knife-like razor edge and one micro-serrated scissor edge. They are identified by dual black finger rings.