ContactUsLinkComponent
Gradle Scissors, OR-Grade, Sklar®
Gradle Scissors, OR-Grade, Sklar®
Catalog # 76193-042
Supplier:  Sklar
undefined
Gradle Scissors, OR-Grade, Sklar®
Catalog # 76193-042
Supplier:  Sklar
Supplier Number:  15-3244
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Supercut™ Gradle Scissors
  • Length:
    3.75"
  • Material:
    Stainless steel
  • Curvature:
    Curved
  • Cat. No.:
    76193-042
  • Blade type:
    Sharp/Sharp
Specifications
About this item
All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
Supercut™Gradle Scissors, similar to Iris scissors, are designed for use in the ophthalmologic, dermatological, and micro-surgery. They combine one knife-like razor edge and one micro-serrated edge. The micro-serrations minimize tearing of tissue so cuts and edges are smooth and clean. With their fine blades and sharp tips, they are good for dissecting and cutting delicate tissue. This instrument is visually identified by one gold finger ring handle.