- Description:Operating Scissors
- Length:7"
- Material:Stainless steel
- Curvature:Curved
- Cat. No.:76191-796
- Blade type:Blunt/Blunt
All Sklar products are industry standard.
- For more than 120 years, Sklar has set the standard for surgical instrumentation
- With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument. These scissors are used for general, multipurpose cutting, and dissecting. Sharp operating scissors cut tougher tissue and are frequently used for dissecting, whereas blunt operating scissors are more often used for cutting materials such as sutures, tubing, or gauze.