ContactUsLinkComponent
Rogge Sterilizing Forceps, OR-Grade, Sklar®
Rogge Sterilizing Forceps, OR-Grade, Sklar®
Catalog # 76191-562
Supplier:  Sklar
undefined
Rogge Sterilizing Forceps, OR-Grade, Sklar®
Catalog # 76191-562
Supplier:  Sklar
Supplier Number:  10-1090
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Rogge Sterilizing Forceps
  • Length:
    9"
  • Material:
    Stainless steel
  • Cat. No.:
    76191-562
Specifications
About this item
All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
Rogge Sterilization Forceps are a versatile utility instrument most commonly used for placing and removing instruments within a sterilization container. The circular, paddle-style tips are serrated so they are useful for grasping a variety of instruments. This product has a non-ratcheted, finger ring handle for easy control. The straight jaws have 1 x 2 prongs tips.