Orders LinkContactUsLinkComponent
Cheatle Sterilization Forceps, OR-Grade, Sklar®
Cheatle Sterilization Forceps, OR-Grade, Sklar®
Cheatle Sterilization Forceps, Stainless steel, 11", Double ('S') Curved
Catalog # 76191-892
Supplier:   Sklar
Supplier #:  10-1011
Cheatle Sterilization Forceps, OR-Grade, Sklar®
Catalog # 76191-892
Supplier:  Sklar
Cheatle Sterilization Forceps, OR-Grade, Sklar®
Cheatle Sterilization Forceps, OR-Grade, Sklar®
Cheatle Sterilization Forceps, Stainless steel, 11", Double ('S') Curved
Price
$178.29
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
Cheatle Sterilization Forceps are a versatile utility instrument most commonly used for placing and removing instruments within a sterilization container. This product has a non-ratcheted finger ring handle for easy control of the instrument.
Specifications
  • Description:
    Cheatle Sterilization Forceps
  • Length:
    11"
  • Material:
    Stainless steel
  • Curvature:
    Double ('S') Curved