Orders LinkContactUsLinkComponent
Actin Polymerization/Depolymerization Assay Kit (Fluorometric), BioVision Inc.
Actin Polymerization/Depolymerization Assay Kit (Fluorometric), BioVision Inc.
Actin Polymerization/Depolymerization Assay Kit, 100 Assays
Catalog # 76265-740
Actin Polymerization/Depolymerization Assay Kit (Fluorometric), BioVision Inc.
Catalog # 76265-740
Supplier:  
CAS Number:  
Actin Polymerization/Depolymerization Assay Kit (Fluorometric), BioVision Inc.
Actin Polymerization/Depolymerization Assay Kit (Fluorometric), BioVision Inc.
Actin Polymerization/Depolymerization Assay Kit, 100 Assays
CAS Number:  
Discontinued
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
The kit to study and quantitate the effect of different compounds, proteins and tissue extracts on actin polymerization and/or depolymerization.
  • Evaluation of critical concentrations of actin polymerization in different conditions
  • The assay is simple, high- throughput compatible and can be completed in less than three hours
Actin, a highly conserved and abundant protein in eukaryotic cells, is one of the major components of cytoskeleton. It can be found as monomeric globular protein, called G actin or it can polymerize into filamentous actin, named F actin.
Actin plays major roles in cell division, cell motility, cell signaling, organelle movement, etc. Mammals have 6 isoforms of Actin, which can be divided into three classes, α, β and γ. Muscle Actin is α class and all other non-muscle actins belong to β and γ classes.
Understanding the effect of different drugs, proteins on actin polymerization and depolymerization is very important for understanding cellular machinery, more importantly because Cytoskeleton is a very important target for cancer therapy.
The kit utilizes a proprietary pyrene-labeled actin molecule that develops a higher fluorescent signal if it undergoes polymerization. The signal can be easily detected using a fluorescence microplate reader.
Specifications
Specifications
  • Description:
    Actin Polymerization/Depolymerization Assay Kit
  • No. of assays/kit:
    100 Assays