Osmolality Standards, Wescor®
About this item
Wescor ampule osmolality standards provide uncompromising accuracy to satisfy the most stringent laboratory quality assurance regimens. Calibration integrity is assured because ampules provide fresh solution with each use
- Ampules provide fresh solution with each use
- Ampule is fitted with a safety breaker sleeve
Delivery information: Each ampule contains 0.4 mL of solution.
3 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Wescor ampule osmolality standards provide uncompromising accuracy to satisfy the most stringent laboratory quality assurance regimens. Calibration integrity is assured because ampules provide fresh solution with each use
- Ampules provide fresh solution with each use
- Ampule is fitted with a safety breaker sleeve
Delivery information: Each ampule contains 0.4 mL of solution.
Documents
Certificates
Search by Lot NumberProduct Family Options
Product Information
- SizeAccuracyAvailabilityPrice
- MSPP-OA-010Osmolality standard, 100 mmol/kg osmolalityOsmolality standard, 100 mmol/kg osmolality60×0.4 ml Ampule±2 mmol/kgLog in for availabilityEach$182.54Supplier Number: OA-010Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:MarketsourceMarketsource
- MSPP-OA-029Osmolality standard, 290 mmol/kg osmolalityOsmolality standard, 290 mmol/kg osmolality60×0.4 ml Ampule±3 mmol/kgLog in for availabilityEach$182.54Supplier Number: OA-029Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:MarketsourceMarketsource
- MSPP-OA-100Osmolality standard, 1000 mmol/kg osmolalityOsmolality standard, 1000 mmol/kg osmolality60×0.4 ml Ampule±5 mmol/kgLog in for availabilityEach$182.54Supplier Number: OA-100Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:MarketsourceMarketsource