Aluminium Sample Pan for Tzero Sample Press Kit, TA Instruments
Sample pan, Aluminum
Catalog # MSPP-901683901
Marketsource
Supplier: TA INSTRUMENTS INC MS
Supplier #: 901683.901
Aluminium Sample Pan for Tzero Sample Press Kit, TA Instruments
Catalog # MSPP-901683901
Supplier: TA INSTRUMENTS INC MS
Marketsource
Jump to:
Product Details & Documents
This aluminum pan is designed for maximum performance in crimp or hermetic sealing of various sample sizes.
- Temperature Range: –180 to 600 °C
Flat bottom design for maximum contact with the Tzero cell sensors.
Specifications
- Description:Sample pan
- Material:Aluminum
Accessories
Product Information
- Catalog #AvailabilityPrice
- Lightweight aluminum lids
Product Information
- Catalog #AvailabilityPrice
- Aluminum hermetic lids
Product Information
- Catalog #AvailabilityPrice
- Hermetic lids with laser-cut vent holeHermetic lids with laser-cut vent holeMSPP-901685901Log in for availabilityPack of 50 - $1,030.77