Single Flow Meter, STEMCELL Technologies
Catalog # MSPP-27311
Supplier: Stemcell Technologies
Specifications
- Description:Single flow meter
- Cat. No.:MSPP-27311
- Supplier no.:27311
Specifications
About this item
Single flow meter is specially designed to control the gas flow in hypoxia incubator chamber to generate a hypoxic environment for tissue culture.
- Calibrated for 5 to 50 L per minute
- Used in mesenchymal stem and progenitor cell research
Maintain the same precision of gas flow as the flow meter used for full size continuous flow carbon dioxide incubator chambers.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').