- Description:Endoscope Basket
- Dimensions:25×6×3"
- Material:Stainless steel/silicone
- Environmentally Preferable:
- Cat. No.:76191-452
All Sklar products are industry standard.
- For more than 120 years, Sklar has set the standard for surgical instrumentation
- With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
Sklars® Endoscope Baskets are designed to store and protect long endoscopic instruments during sterilization. This basket has four top and bottom silicone inserts to support the instruments as well as a lens basket. This product comes with a removable lid with secure fasteners.