Order Entry
United States
ContactUsLinkComponent
Blunt-End Needles, 16 Gauge, STEMCELL Technologies
Blunt-End Needles, 16 Gauge, STEMCELL Technologies
Catalog # MSPP-28110
undefined
Blunt-End Needles, 16 Gauge, STEMCELL Technologies
Catalog # MSPP-28110
Supplier Number:  28110
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Blunt-end Needle, Sterile
  • Length:
    38.1 mm (1.50")
  • Environmentally Preferable:
  • Cat. No.:
    MSPP-28110
  • Gauge:
    16
  • Supplier no.:
    28110

Specifications

About this item

Ideal for safe and accurate handling of methylcellulose-based media. Blunt end reduces the risk of needle stick injuries.

  • With a metal Luer lock hub

Manufactured from non-pyrogenic, non-toxic materials.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').