Order Entry
Puerto Rico
ContactUsLinkComponent
 
Plasmocin™ Treatment - Mycoplasma Elimination Reagent, InvivoGen
Catalog # MSPP-ANTMPT
Supplier:  Invivogen
CAS Number:  
Plasmocin™ Treatment - Mycoplasma Elimination Reagent, InvivoGen
Catalog # MSPP-ANTMPT
Supplier:  Invivogen
Supplier Number:  ANT-MPT
CAS Number:  

Specifications

  • Description:
    Plasmocin™ Treatment
  • Cat. No.:
    MSPP-ANTMPT
  • Supplier no.:
    ANT-MPT

Specifications

About this item

Plasmocin™ treatment used for the elimination of mycoplasma contamination in cell cultures without affecting cell stemness, pluripotency or viability.

  • Endotoxin level: <2 EU/mg
  • Store at –20 °C

Plasmocin™ is active on both extracellular and intracellular mycoplasmas. It is active at low concentrations on a broad range of gram positive and gram negative bacteria that are otherwise resistant to the mixture of streptomycin and penicillin, and exhibits no toxicity towards eukaryotic cells.

One 1 ml vial is sufficient for 660 ml to 2 liters of culture.

Delivery: Supplied as a cell culture tested, sterile filtered yellow solution at 25 mg/ml.
Caution: For research use only. Not for human or veterinary use.