Order Entry
Puerto Rico
ContactUsLinkComponent
EnzyChrom™ D-Mannitol Assay Kit, BioAssay Systems
EnzyChrom™ D-Mannitol Assay Kit, BioAssay Systems
Catalog # 76236-140
Supplier:  BIOASSAY SYSTEMS
CAS Number:  
undefined
EnzyChrom™ D-Mannitol Assay Kit, BioAssay Systems
Catalog # 76236-140
Supplier:  BIOASSAY SYSTEMS
Supplier Number:  EMNT-100
CAS Number:  

Specifications

  • Description:
    EnzyChrom™ D-Mannitol Assay Kit
  • Size:
    100 assays
  • Cat. No.:
    76236-140

Specifications

About this item

For quantitative determination of D-mannitol in biological, food, beverage and agriculture samples.

  • Fast and sensitive, convenient
  • Use of 20 μl sample
  • Linear detection range 0.007 to 3 mM mannitol in 96-well plate assay
  • Room temperature assay, no 37 °C heater is needed
  • High-throughput, "Add-mix-read" type assay
  • Can be readily automated as a high-throughput 96-well plate assay for thousands of samples per day
  • Method: OD565 nm

The procedure involves adding a single working reagent, and reading the absorbance after 30 minutes. D-Mannitol (C₆H₁₄O₆) is a sugar alcohol used in dietary supplement, sweetener, intestinal permeability test for leaky gut, etc. It also serves as a coating for hard candies, dried fruits, and chewing gums due to its low ability to attract and hold water molecules. In addition, it is an osmoprotectant for plants and is used clinically in osmotherapy to reduce intracranial pressure.

D-mannitol assay kit is based on mannitol dehydrogenase catalyzed oxidation of D-mannitol, which generates D-fructose and NADH that reduces a formazan (MTT) dye. The intensity of product color, measured at 565 nm is directly proportional to D-mannitol concentration in the sample. Samples: Biological, food, beverage and agricultural products.