Orders LinkContactUsLinkComponent
Lymphoprep™ Density Media
Lymphoprep™ Density Media
Marketsource
Lymphoprep™ Density Media
Marketsource
Certificates
Lymphoprep™ Density Media
Lymphoprep™ Density Media
4 Options Available Below
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Reliably isolate mononuclear cells from peripheral blood, cord blood, or bone marrow with Lymphoprep™, a cost-effective alternative to Ficoll-Paque™.
  • Density gradient medium for the isolation of mononuclear cells
  • Rapid, simple and reliable
  • Provides excellent results with blood samples from most normal individuals and patients
Substitute Lymphoprep™ for Ficoll-Paque™ without changing existing protocols to achieve similar cell purity and recovery rates. This medium is fully compatible with both SepMate™ and RosetteSep™. Formulated with sodium diatrizoate (9.1% w/v) and polysaccharide (5.7% w/v), Lymphoprep™ has a density of 1.077 g/ml.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Product Family Options
Product Information
  • Availability
    Price
  • MSPP-07801
    Lymphoprep™ density gradient medium, 250 ml bottle
    Lymphoprep™ density gradient medium, 250 ml bottle
    Log in for availability
    Log in for availability
    Each
    Each - $168.08
    $168.08
    Each
       $168.08
    Minimum order quantity is 1
    Supplier Number: 18060
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    Marketsource
  • MSPP-7811
    Lymphoprep™ density gradient medium, 4×250 ml bottles
    Lymphoprep™ density gradient medium, 4×250 ml bottles
    Log in for availability
    Log in for availability
    Pack of 4
    Pack of 4 - $447.23
    $447.23
    Pack of 4
       $447.23
    Minimum order quantity is 1
    Supplier Number: 07811
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    Marketsource
  • MSPP-07851
    Lymphoprep™ density gradient medium, 500 ml bottle
    Lymphoprep™ density gradient medium, 500 ml bottle
    Log in for availability
    Log in for availability
    Each
    Each - $273.31
    $273.31
    Each
       $273.31
    Minimum order quantity is 1
    Supplier Number: 18061
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    Marketsource
  • MSPP-07861
    Lymphoprep™ density gradient medium, 6×500 ml bottles
    Lymphoprep™ density gradient medium, 6×500 ml bottles
    Log in for availability
    Log in for availability
    Each
    Each - $1,001.15
    $1,001.15
    Each
       $1,001.15
    Minimum order quantity is 1
    Supplier Number: 07861
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    Marketsource