ContactUsLinkComponent
Tryptic Soy Agar Plates with 5% Sheep Blood, General Laboratory Products
Tryptic Soy Agar Plates with 5% Sheep Blood, General Laboratory Products
Catalog # 101205-690
Minority or Woman-Owned Business Enterprise
CAS Number:  
Tryptic Soy Agar Plates with 5% Sheep Blood, General Laboratory Products
Catalog # 101205-690
Supplier Number:  MP7139
Minority or Woman-Owned Business Enterprise
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Tryptic Soy Agar Monoplate, 10 per Pack
  • Cat. No.:
    101205-690
  • Plate size:
    100×15 mm
  • Supplier no.:
    M1006
Specifications
About this item
Tryptic soy agar with 5% sheep blood is used for cultivating fastidious microorganisms and for the visualization of hemolytic reactions produced by many bacterial species.