Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Dextrose solution 10% USP for injection

Dextrose solution 10% USP for injection

Plastic container, Viaflex®, 500 ml
Catalog # 68000-167
Supplier:   Baxter
Supplier #:  2B0163Q
Dextrose solution 10% USP for injection
Catalog # 68000-167
Supplier:  Baxter
CAS Number:  50-99-7
Dextrose solution 10% USP for injection
Dextrose solution 10% USP for injection

Plastic container, Viaflex®, 500 ml

CAS Number:  50-99-7
Price
$914.02
Log in for availability
Case of 24 (500ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
  • Total calories: 340 kcal/l
  • Specific gravity: 1.03
  • Fill range volume (ml): 1030 to 1070
  • Store at room temperature (25 °C)
  • mOsmol/L (calc): 505
  • NDC Number: 0338-0023-03
  • pH 4

 : USP specified.

Synonyms:  Corn sugar, ᴅ-(+)-Glucose, Glucose, D-(+)-Dextrose, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, Glucolin, Dextrose, D-Glucose
  • CAS Number:50-99-7
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00148912
  • Molecular Weight:180.16 g/mol
Specifications

Specifications

  • Pack type:
    Plastic container, Viaflex®
  • Size:
    500 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.