Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse Recombinant CCL21 (from E. coli)
Mouse Recombinant CCL21 (from E. coli)
Supplier:  Biolegend
Certificates
undefined
Mouse Recombinant CCL21 (from E. coli)
Supplier:  Biolegend

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <0.01 ng/ug of protein
  • Gene ID:
    18829
  • Protein Synonyms:
    SCYA21|6CKine|Exudos 2|Secondary lymphoid tissue chemokine (SLC)
  • Protein/Peptide Name:
    CCL21
  • Purity:
    >98%
  • Molecular Weight:
    12 kDa
  • Formulation:
    0.22 µm filtered protein solution is in PBS
  • Tested Applications:
    Bioassay

Specifications

Frequently Bought Together