- Description:Unna-Vidal Comedone Extractor
- Length:11.4 cm (4½")
- Material:Stainless steel
- Curvature:Straight
- Cat. No.:76193-084
Sklar's® Unna-Vidal Comedone Extractor is a three piece dermal instrument used to treat facial blemishes. This double-ended instrument has one round end with a small fenestration specifically designed for extraction and one sharp, pointed probe end. The tips are replaceable.
- For more than 120 years, Sklar has set the standard for surgical instrumentation
- With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure