Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Ethidium bromide, dark red solution for molecular biology

Ethidium bromide, dark red solution for molecular biology

Glass bottle, amber, 10 ml
Catalog # IC11ETBC1001
Toxic
Health
Supplier:   MP Biomedicals
Supplier #:  11ETBC1001
Ethidium bromide, dark red solution for molecular biology
Catalog # IC11ETBC1001
Supplier:  MP Biomedicals
CAS Number:  1239-45-8
Ethidium bromide, dark red solution for molecular biology
Ethidium bromide, dark red solution for molecular biology

Glass bottle, amber, 10 ml

CAS Number:  1239-45-8
Price
$78.00
Log in for availability
Each (10ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

A 10 mg/mL Ready-To-Use solution of EtBr in specially filtered, deionized water.

  • Grade : Molecular Biology
  • Concentration : (10 mg/ml)
  • Suitability : Suitable for gel electrophoresis
  • Key Applications : Nucleic acid stain for PAGE or agarose gel electrophoresis; Used in fluorimetric assays for nucleic acids
  • Application Areas : Nucleic Acid Electrophoresis; Blotting & Detection; Nucleic Acid Gel Electrophoresis & Blotting
  • Product Type : Biochemicals
  • Analyte(s) Detected : dsDNA; dsRNA
  • Biochemical Category : Labels, Dyes & Stains
  • Chemical Class : Labels, Dyes & Stains
  • Vapor Pressure : 1.2 X 10-12 mm Hg at 25 °C
  • UV/Visible Absorbance : 254 nm
  • Presentation : Dark Red Solution
  • Detection Method : UV gel box (254 nm)
  • Solubility : Soluble in water.
  • Recommended Replacement For : Ethidium Bromide Powder
  • Certifications : DNase, RNase Protease Free
  • Detection/Incubation Time : 15-30 min with 0.5 mg/mL solution

Storage: protect from light.

Synonyms:  3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide, 3,8-Diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide
  • CAS Number:1239-45-8
  • Formula:C₂₁H₂₀BrN₃
  • MDL Number:MFCD00011724
  • Molecular Weight:394.31 g/mol
  • Storage Temperature:Refrigerator
  • UN:2810
  • ADR:,III
Specifications

Specification Test Results

  • Identity Test:
    Passes

Specifications

  • Pack type:
    Glass bottle, amber
  • Size:
    10 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.