Order Entry
Puerto Rico
ContactUsLinkComponent
Fluorescein *Fluorescence Reference Standard*, Anaspec Inc.
Fluorescein *Fluorescence Reference Standard*, Anaspec Inc.
Catalog # 103010-702
Supplier:  Anaspec Inc
CAS Number:  
Fluorescein *Fluorescence Reference Standard*, Anaspec Inc.
Catalog # 103010-702
Supplier:  Anaspec Inc
Supplier Number:  AS-80001
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Fluorescein
  • Size:
    1 g
  • Storage Conditions:
    -20°C desiccated and protected from light
  • Shipping Conditions:
    RT
  • Cat. No.:
    103010-702
  • Format:
    Lyophilized
  • Absorbency:
    Abs/Em = 490/514 nm
  • Supplier no.:
    AS-80001

Specifications

About this item

pH-dependent fluorescence; Maximum intensity observed in basic solutions.