Order Entry
Puerto Rico
ContactUsLinkComponent
Medium 199 (E-199) without L-Glutamine and Phenol Red, Quality Biological
Medium 199 (E-199) without L-Glutamine and Phenol Red, Quality Biological
Catalog # 76266-202
CAS Number:  
undefined
Medium 199 (E-199) without L-Glutamine and Phenol Red, Quality Biological
Catalog # 76266-202
Supplier Number:  112-254-101
CAS Number:  

Specifications

  • Description:
    E-199
  • Sterility:
    Sterile filtered
  • Shipping Conditions:
    Ambient temperature
  • Osmolality:
    290 ±5% mOsm/Kg H₂0
  • Cat. No.:
    76266-202
  • Shelf life:
    24 months

Specifications

About this item

E199 or Medium 199 with Earle’s Salt is one of the best known media in the history of cell culture. Morgan, Morton and Parker’s Medium 199 was the first really successful attempt to develop a totally defined medium. Medium 199 was originally developed for nutritional studies of chick embryo fibroblasts.

  • Without Phenol Red
  • Without L-glutamine, Hepes

E199 has broad species applicability, particularly for cultivation of non-transformed cells. Compared to other basal media, Medium 199 contains unique components, including adenine, adenosine, hypoxanthine, thymine, and additional vitamins. Medium 199 is available with Earle’s salts for use in a CO2 incubator, or with Hanks’ salts for use without CO2.

E199 has been used for many different types of cells, especially non-transformed cells, and proved particularly useful in polio vaccine production and other virology applications such as vaccine production and in vitro cultivation of primary explants of mouse pancreatic epithelium, and rat lens tissues. QBI offers a variety of E-199 Medium modifications for a range of cell culture applications.

Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Caution: Research Use Only