Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Size:1 mg
- Storage Conditions:–20 °C desiccated and protected from light
- Protein/Peptide Name:Collagen (Type I) (soluble)
- Solubility:Water soluble
- Shipping Temperature:Shipped in Blue Ice
- Cat. No.:103011-292
- Supplier no.:AS-85111
Specifications
About this item
AnaSpec offers two types of fluoresceinated collagen (type I). This product is water soluble and is used for quick fluorometric measurement of collagenase activity. In this protease substrate, collagen is heavily labeled with FITC, resulting in almost total quenching of the conjugate's fluorescence. Protease-catalyzed hydrolysis relieves this quenching conjugate, yielding brightly green fluorescent dye-labeled peptides. The increase in fluorescence intensity is directly proportional to protease activity. This fluoresceinated collagen is useful for detecting MMP-1 activity.
- Absorbance: Abs/Em = 492/515 nm