Order Entry
Puerto Rico
ContactUsLinkComponent
SMCC Activated B - PE
SMCC Activated B - PE
  103010-440
 :  
SMCC Activated B - PE
  103010-440
 :  AS-72109
 :  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Size:
    1 mg
  • Storage Conditions:
    4 °C, Do NOT freeze!
  • Protein/Peptide Name:
    SMCC Activated B - PE
  • Shipping Temperature:
    Shipped in Blue Ice
  • Cat. No.:
    103010-440
  • Supplier no.:
    AS-72109

 

 

B-Phycoerythrin (B-PE), a fluorescent protein from phycobiliprotein family, is isolated from cyanobacteria and eukaryotic algae. SMCC activated B-PE is chemically modified with SMCC. SMCC reacts with the primary amine on B-PE and introduces maleimide groups to B-PE. These maleimide groups easily react with thiol groups of target protein without the need for any additional activation, resulting in convenient conjugation of B-PE with proteins. Its primary absorption peak is at 545 nm with secondary peak at 563 nm. B-PE and the closely related R-PE are the most intensely fluorescent phycobiliproteins having orange fluorescence. They are significantly brighter and more photostable than conventional organic fluorophores. B-PE conjugates have been widely used in applications such as flow cytometry and multi-color immunofluorescent staining.

  • Absorbance: Abs/Em = 545/575 nm