Order Entry
Puerto Rico
ContactUsLinkComponent
AnaTag™ B-PE Labeling Kit, Anaspec Inc.
AnaTag™ B-PE Labeling Kit, Anaspec Inc.
Catalog # 103010-182
Supplier:  Anaspec
CAS Number:  
AnaTag™ B-PE Labeling Kit, Anaspec Inc.
Catalog # 103010-182
Supplier:  Anaspec
Supplier Number:  AS-72112
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    AnaTag™ B-PE Labeling Kit
  • Size:
    1 kit
  • Storage Conditions:
    4°C
  • Shipping Conditions:
    20
  • Cat. No.:
    103010-182
  • Abs/Em:
    545,563/578 nm
  • No. of reactions:
    1 labeling reaction (1x1 mg IgG)
  • Supplier no.:
    AS-72112

Specifications

About this item

The AnaTag™ B-PE Labeling Kit is optimized for use in the conjugation of B-Phycoerythrin (B-PE) to antibodies. B-PE, a fluorescent protein from phycobiliprotein family, has a primary absorption peak at 545 nm with a secondary peak at 563 nm and maximum emission at 578 nm.

B-PE and the closely related R-PE are the most intensely fluorescent phycobiliproteins having orange fluorescence. They are significantly brighter and more photostable than conventional organic fluorophores. B-PE labeled primary and secondary antibodies are used in applications such as flow cytometry and multi-color immunofluorescent staining. The AnaTagTM B-PE Labeling Kit contains SH-reactive B-PE. SMCC modified B-PE reacts with the thiol groups of target antibody without the need for additional activation, thus simplifying the conjugation protocol. The amount of B-PE supplied in this kit is sufficient for labeling up to 1 mg of antibody. The kit provides all reagents, purification columns and a detailed step-by-step protocol. Native and SMCC-activated B-PE are also available as separate products.