To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Host:
- Description:AnaTag™ Biotin Protein Labeling Kit
- Size:1 kit
- Environmentally Preferable:
- Storage Conditions:4°C
- Shipping Conditions:20
- Cat. No.:103010-370
- No. of reactions:3 labeling reactions (3× 10 mg protein)
- Supplier no.:AS-72057
Specifications
About this item
Biotin, SE is amino-reactive biotinylating reagent for peptides and proteins; it has a better avidin-binding affinity than biotin. Biotin, SE (d-Biotin-N-hydroxysuccinimide ester) contains a five-carbon spacer arm that reduces steric hindrance associated with binding four biotinylated molecules per one avidin and results in enhanced detection sensitivity.
The succinimidyl ester (SE) reactive form of biotin reacts with the amine group on the protein and forms stable carboxamide bonds, which is identical to natural peptide bonds. Biotin-protein conjugates are very stable and can withstand the conditions of immunofluorescent staining, fluorescence in situ hybridization, flow cytometry and other biological applications without hydrolysis. The kit has all essential components for performing the conjugation reaction and for purifying the biotin-protein conjugates. The kit is able to perform three conjugation reactions with each reaction labeling up to 10 mg IgG.