Order Entry
Puerto Rico
ContactUsLinkComponent
AnaTag™ Biotin Microscale Protein Labeling Kit, Anaspec Inc.
AnaTag™ Biotin Microscale Protein Labeling Kit, Anaspec Inc.
  103010-372
 :  Anaspec
 :  
AnaTag™ Biotin Microscale Protein Labeling Kit, Anaspec Inc.
  103010-372
 :  Anaspec
 :  AS-72058
 :  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Description:
    Biotin Microscale Protein Labeling Kit AnaTag™
  • Size:
    1 kit
  • Storage Conditions:
    4°C
  • Shipping Conditions:
    20
  • Cat. No.:
    103010-372
  • Supplier no.:
    AS-72058

 

 

Biotin, SE is amino-reactive biotinylating reagent for peptides and proteins; it has a better avidin-binding affinity than biotin. Biotin, SE (d-Biotin-N-hydroxysuccinimide ester, Biotin, NHS ester) contains a five-carbon spacer arm that reduces steric hindrance associated with binding four biotinylated molecules per one avidin and results in enhanced detection sensitivity.

The succinimidyl ester (SE) reactive form of biotin reacts with the amine group on the protein and forms stable carboxamide bonds, which is identical to natural peptide bonds. Biotin-protein conjugates are very stable and can withstand the conditions of immunofluorescent staining, fluorescence in situ hybridization, flow cytometry and other biological applications without hydrolysis. The kit has all essential components for performing the conjugation reaction and for purifying the biotin-protein conjugates. The kit is able to perform three conjugation reactions with each reaction labeling up to 200 µg IgG. Kit Size: 3 labeling reactions (3 x 200 µg protein)