Order Entry
Puerto Rico
ContactUsLinkComponent
 
AnaTag™ 5 - TAMRA Protein Labeling Kit *Ultra Convenient*, Anaspec
  103010-382
 :  Anaspec
 :  
AnaTag™ 5 - TAMRA Protein Labeling Kit *Ultra Convenient*, Anaspec
  103010-382
 :  Anaspec
 :  AS-72063
 :  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Description:
    5 - TAMRA Protein Labeling Kit AnaTag™ 'Ultra Convenient'
  • Size:
    1 kit
  • Storage Conditions:
    4°C
  • Shipping Conditions:
    20
  • Cat. No.:
    103010-382
  • Abs/Em:
    547/574 nm
  • Supplier no.:
    AS-72063

 

 

TAMRA is one of the most popular fluorophores used in various bioconjugations. 5-TAMRA is the purified single isomer of 5(6)-TAMRA. It is preferred for some complicated biological applications where reproducibility is more critical than material cost since the minor positional difference between 5-TAMRA and 6-TAMRA might affect some biological properties of the underlying conjugates.

5-TAMRA is more often used than 6-TAMRA for labeling peptides and proteins. 6-TAMRA is predominately used for labeling nucleotides and sequencing nucleic acids. The 5-TAMRA-protein conjugates are suitable for immunofluorescent staining, in situ hybridization, flow cytometry and other biological applications. The kit has all essential components for performing the conjugation reaction and for purifying the 5-TAMRA-protein conjugates. The kit is able to perform three conjugation reactions with each reaction labeling up to 5 mg IgG. Kit Size: 3 labeling reactions (3 x 5 mg protein)