Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle, amber
- CAS No.:9001-63-2
- Source:Egg white
- Species:Chicken
- Size:1 g
- Storage Conditions:2...8 °C
- MDL No.:MFCD00131557
- Enzyme Name:Lysozyme Type VI
- Enzyme Activity:≥23500 Shugar U/mg
- Purity:≥95%
- pH:3.0 - 4.5
- Cat. No.:76177-422
Specifications
About this item
Lysozyme (muramidase) hydrolyzes preferentially the b-1,4 glucosidic linkages between N-acetylmuramic acid and N-acetylglucosamine which occur in the mucopeptide cell wall structure of certain microorganisms, such as Micrococcus lysodeikticus.
- Grade : Molecular Biology Reagent
- Specific Activity : ≥23500 Shugar units/mg
- Key Applications : Cell Lysis
- Product Type : Proteins, Enzymes & Peptides
- Biochemical Category : Proteins, Enzymes & Peptides
- Extinction Coefficient : (E1%) 26.4 (280 nm) (Lit.)
- Presentation : White to Off-white Powder
- Format Type : VI, 3X Crystallized, Salt-Free, Albumin-Free, Lyophilized
- Isoelectric point : (pI) pH 11.0 (lit)
- pH : 3.6 (Lit)
- NOTES : Chloride ≤3.5%