Order Entry
Puerto Rico
ContactUsLinkComponent
Bovine Serum Albumin (BSA), Fraction V, MP Biomedicals
Bovine Serum Albumin (BSA), Fraction V, MP Biomedicals
Supplier:  MP Biomedicals
Certificates
Bovine Serum Albumin (BSA), Fraction V, MP Biomedicals
Supplier:  MP Biomedicals

Specifications

  • CAS No.:
    9048-46-8
  • Species:
    Bovine
  • Storage Conditions:
    +4 °C
  • Endotoxin Content:
    <2.0 EU/mg
  • Protein/Peptide Name:
    Albumin
  • Endotoxin Level:
    Low
  • pH:
    6.8-7.2

Specifications