Order Entry
Puerto Rico
ContactUsLinkComponent
Bovine Aprotinin (from Lung), MP Biomedicals
Bovine Aprotinin (from Lung), MP Biomedicals
Supplier:  MP Biomedicals
Certificates
undefined
Bovine Aprotinin (from Lung), MP Biomedicals
Supplier:  MP Biomedicals

Specifications

  • CAS No.:
    9087-70-1
  • Source:
    Lung
  • Species:
    Bovine
  • Storage Conditions:
    Store at +4 °C.
  • Form:
    white powder
  • MDL No.:
    MFCD00130541
  • Enzyme Name:
    Aprotinin
  • Enzyme Activity:
    ≥3 TIU/mg
  • pH:
    5.0 - 7.0

Specifications

Frequently Bought Together