Bovine Ribonuclease A (from Pancreas), MP Biomedicals
Supplier: MP Biomedicals
Certificates
About this item
Ribonuclease belongs to pancreatic ribonuclease family. RNase A is an endoribonuclease that attacks at the 3¢ phosphate of a pyrimidine nucleotide. The sequence of pG-pG-pC-pA-pG will be cleaved to give pG-pG-pCp and A-pG.
- Grade : Molecular Biology Reagent
- Specific Activity : ≥70 Kunitz units/mg
- Key Applications : Purification of DNA; Analysis of RNA sequences; RNase protection assays; Hydrolyze RNA contained in protein samplesApplication Areas
- Product Type : Proteins, Enzymes & Peptides
- Extinction Coefficient : (E1%) E1%280 = ~7.3
- Presentation : White Powder
- Format : Lyophilized powder, Salt-free and protease-free
- Isoelectric point (pI) : 9.45
- pH Optimum pH: 7.0 to 7.5
- NOTES : Inhibitors: Ribonuclease is inhibited by heavy metal ions and is competitively inhibited by DNA. The effect of denatured DNA is much greater than that of the native nucleic acid. Composition: The composition and structure of ribonuclease A has been extensively investigated. The tertiary structure indicates that flexibility in a three-dimensional structure is essential for optimal catalytic activity.
- Specificity : The enzyme is specific for pyrimidine nucleoside linkages, i.e., those cleaving cyclic 2',3'-pyrimidine nucleotide residues either singly or at the termination of purine nucleotide chains.
- Solubility: Dissolves readily at 2 mg/ml in analytical grade water to give a clear colorless solution.
Specifications
- CAS No.:9001-99-4
- Source:Pancreas
- Species:Bovine
- Storage Conditions:−20 °C
- Form:White powder
- MDL No.:MFCD00132181
- Enzyme Name:Ribonuclease A
- Enzyme Activity:≥70 Kunitz units/mg
Specifications
Frequently Bought Together