Order Entry
Puerto Rico
ContactUsLinkComponent
Pig Pancreatin (from Pancreas), MP Biomedicals
Pig Pancreatin (from Pancreas), MP Biomedicals
Supplier:  MP Biomedicals
Certificates
Pig Pancreatin (from Pancreas), MP Biomedicals
Supplier:  MP Biomedicals
Certificates

Specifications

  • CAS No.:
    8049-47-6
  • Source:
    Pancreas
  • Species:
    Pig
  • Storage Conditions:
    −20 °C
  • Form:
    Cream Powder
  • MDL No.:
    MFCD00131789
  • Enzyme Name:
    Pancreatin
  • Enzyme Activity:
    Lipase Activity

Specifications

Related Information