Order Entry
Puerto Rico
ContactUsLinkComponent
AnaTag™ HiLyte™ Fluor 555 Protein Labeling Kit *Ultra Convenient* [3 labeling reactions (3 x 5 mg protein)], Anaspec
AnaTag™ HiLyte™ Fluor 555 Protein Labeling Kit *Ultra Convenient* [3 labeling reactions (3 x 5 mg protein)], Anaspec
Catalog # 103010-350
Supplier:  Anaspec
CAS Number:  
AnaTag™ HiLyte™ Fluor 555 Protein Labeling Kit *Ultra Convenient* [3 labeling reactions (3 x 5 mg protein)], Anaspec
Catalog # 103010-350
Supplier:  Anaspec
Supplier Number:  AS-72045
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Fluor 555 Protein Labeling Kit [3 labeling Reactions (3 x 5 mg Protein)] AnaTag™ HiLyte™ 'Ultra Convenient'
  • Size:
    1 kit
  • Storage Conditions:
    4°C
  • Shipping Conditions:
    20
  • Cat. No.:
    103010-350
  • Abs/Em:
    553/568 nm
  • Supplier no.:
    AS-72045

Specifications

About this item

HiLyte™ Fluor 555 acid, SE is an excellent amine-reactive fluorescent labeling dye that generates the protein conjugates that are only slightly red-shifted compared to those of Cy™ 3 dyes, resulting in an optimal match to filters designed for CyTM dyes.

The HiLyte™ Fluor 555-protein conjugates are very stable and suitable for immunofluorescent staining, in situ hybridization, flow cytometry and other biological applications. Additionally, our in-house data indicated that HiLyte™ Fluor 555 is superior to CyTM 3 in fluorescence polarization-based assays. The kit has all essential components for performing the conjugation reaction and for purifying the HiLyte™ Fluor 555-protein conjugate. The entire process takes only half an hour. The kit is able to perform three conjugation reactions with each reaction labeling up to 5 mg IgG. Kit Size: 3 labeling reactions (3 x 5 mg protein)