ContactUsLinkComponent
Moeltgen Goniometer, Sklar®
Moeltgen Goniometer, Sklar®
Catalog # 76193-060
Supplier:  Sklar
undefined
Moeltgen Goniometer, Sklar®
Catalog # 76193-060
Supplier:  Sklar
Supplier Number:  06-2821
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Moeltgen Goniometer
  • Length:
    9½"
  • Material:
    Stainless steel
  • Cat. No.:
    76193-060
Specifications
About this item
All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
Sklar's® Moeltgen Goniometer is used to measure range of motion, primarily in occupational and physical therapy.