Specifications
- Color:Black
- Description:Stethoscope
- Material:Stainless steel/PVC
- Cat. No.:76191-568
- Type:Cardiology
Specifications
About this item
All Sklar products are industry standard.
- For more than 120 years, Sklar has set the standard for surgical instrumentation
- With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
Sklar's® Dual Head Cardiology Stethoscope allows the user to hear high, medium, and low frequency sounds, specifically those identified with the heart such as blood flow, clicks, and murmurs. The cardiology stethoscope has thicker, shorter PVC tubing than a standard stethoscope. This provides a shorter distance for the sounds to travel and decreases interference of curled tubing that can occur with longer stethoscopes. The result is a more accurate identification of what is being heard. The Sklar Dual Head Cardiology Stethoscope features a stainless steel diaphragm and bell to further improve sound transmission. This stethoscope measures 25" and is available in a variety of tubing colors.
The Sklar Dual Head Cardiology Stethoscope features a stainless-steel diaphragm and bell to further improve sound transmission. This stethoscope measures 25" and is available in a variety of tubing colors.