To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Camelid VHH
- Antigen Symbol:Camelid VHH
- Clonality:Monoclonal
- Clone:96A3F5
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Camel,Llama,Alpaca
- Western Blot:Yes
- Size:200 μl
- Cross Adsorption:No
- Form:Liquid
- Storage Buffer:PBS, pH 7.4, 10 mg/ml BSA, 0.01% thimerosal and 50% glycerol
- Storage Temperature:Store at -20 °C. This product is stable for 1 year upon receipt, when handled and stored as instructed
- Concentration:0.5 mg/ml
- Shipping Temperature:Ice
- Immunogen:Llama sdAb
- Purification:Protein A affinity column
- Cat. No.:76197-318
- Packaging:Vial
Specifications
About this item
GenScript MonoRab™ Rabbit Anti-Camelid VHH Antibody [HRP], mAb is specific for Camelid sdAb (Alpaca, Llama and Camel). The monoclonal antibody has no cross-reactivity with mouse, rat, rabbit, chicken, goat or human immunoglobulins.
- Anti-VHH antibodies
- MonoRab™ technology guarantees high affinity: Kd ≈ 10-11 M
- No cross reactivity with other species
- Specific to camelid IgG2 and IgG3
- Recognize conformational epitopes of the VHH domain
- Recognize both camelid and humanized VHHs
GenScript has developed a series of rabbit monoclonal (MonoRab ) anti-VHH antibodies to facilitate your research of VHHs. Our anti-VHH antibodies recognize conformational epitopes of the VHH domain of camelid lgG2 & lgG3 subclasses. It is not necessary to add a tag to VHH for detection anymore.
GenScript MonoRabᵀᴹ Rabbit Anti-Camelid VHH Antibody [HRP], mAb is produced from a hybridoma resulting from the fusion of partner and B-lymphocytes obtained from a rabbit immunized with llama sdAb.
GenScript MonoRabᵀᴹ Rabbit Anti-Camelid VHH Antibody [HRP], mAb is specific for Camelid sdAb (Llama and Camel). The monoclonal antibody has no cross-reactivity with mouse, rat, rabbit, chicken, goat or human immunoglobulins.
Type: Secondary
Antigen: Camelid VHH
Clonality: Monoclonal
Clone: 96A3F5
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Llama