Helix-IN, Transfection Reagent, OZ Biosciences
Supplier: OZ Biosciences
Certificates
About this item
OZ Biosciences revolutionizes Polyfection with the design of Helix-IN™, a novel patented Cationic Hydroxylated Amphiphilic Multi-block Polymer (CHAMP™ Technology). This novel bi-functional co-polymer is biocompatible, ionizable, pH responsive and biodegradable.
- Broad Spectrum Transfection Reagent for cell lines and hard-to-transfect cells
- High transfection efficiency & increased transgene expression even with low DNA amount
- High intracellular protein production while preserving viability
- High secreted protein production while minimizing cellular stress
- Biodegradable-Avoid secondary effects
- Compatible with any culture medium
Delivery: Shipped at room temperature.
Packaging: Store at -20°C