Orders LinkContactUsLinkComponent
ALLN
ALLN
10 mg
Catalog # 95043-376
Supplier:   G-Biosciences
Supplier #:  786-057
ALLN
Catalog # 95043-376
Supplier:  G-Biosciences
CAS Number:  
ALLN
CAS Number:  
Price
$416.73
Log in for availability
Each (10mg)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
A selection of individual protease inhibitors are offered for researchers who wish to design their own cocktails, supplement existing cocktails, or screen for specific proteases
Also available is the Protease Inhibitor Set, which contains all 12 of our individual protease inhibitors, each in a ready-to-use solution at a 100X concentration. The 1X concentration of the protease inhibitors is designed to give >90% inhibition in crude tissue extracts. Various concentrations and/or combinations of protease inhibitors may be used to inhibit a broad spectrum of protease activity.
G-Biosciences offers the following individual protease inhibitors as a dry powder: AEBSF, ALLN, Antipain dihydrochloride, Aprotinin, Bestatin, Chymostatin, E-64, EDTA-Na2, Leupeptin, Pepstatin, Phosphoramidon, and PMSF.
Specifications
Specifications
  • Size:
    10 mg
  • Cat. No.:
    95043-376
  • Protein/Peptide Name:
    ALLN