ContactUsLinkComponent
Chymostatin
Chymostatin
Catalog # 82022-382
Supplier:  G-Biosciences
CAS Number:  
undefined
Chymostatin
Catalog # 82022-382
Supplier:  G-Biosciences
Supplier Number:  786-048
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Size:
    5 mg
  • Protein/Peptide Name:
    Chymostatin
  • Cat. No.:
    82022-382
  • Supplier no.:
    786-048
Specifications
About this item
A selection of individual protease inhibitors are offered for researchers who wish to design their own cocktails, supplement existing cocktails, or screen for specific proteases
  • Inhibitor of alpha, beta, gamma, and delta chymotrypsins.
Also available is the Protease Inhibitor Set, which contains all 12 of our individual protease inhibitors, each in a ready-to-use solution at a 100X concentration. The 1X concentration of the protease inhibitors is designed to give >90% inhibition in crude tissue extracts. Various concentrations and/or combinations of protease inhibitors may be used to inhibit a broad spectrum of protease activity.
G-Biosciences offers the following individual protease inhibitors as a dry powder: AEBSF, ALLN, Antipain dihydrochloride, Aprotinin, Bestatin, Chymostatin, E-64, EDTA-Na2, Leupeptin, Pepstatin, Phosphoramidon, and PMSF.